missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BPHL Protein
A DNA sequence encoding the Homo sapiens (Human) BPHL, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7794-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001289706.1 | |
BPHL Protein | |
Research Use Only | |
BPHL | |
Recombinant Protein | |
E. coli | |
MPRNLLYSLL SSHLSPHFST SVTSAKVAVN GVQLHYQQTG EGDHAVLLLP GMLGSGETDF GPQLKNLNKK LFTVVAWDPR GYGHSRPPDR DFPADFFERD AKDAVDLMKA LKFKKVSLLG WSDGGITALI AAAKYPSYIH KMVIWGANAY VTDEDSMIYE GIRDVSKWSE RTRKPLEALY GYDYFARTCE KWVDGIRQFK HLPDGNICRH LLPRVQCPAL IVHGEKDPLV PRFHADFIHK HVKGSRLHLM PEGKHNLHLR FADEFNKLAE DFLQ | |
Greater than 90% as determined by SDS-PAGE. |
670 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |