missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CAS1 domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) CAS1 domain-containing protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7946-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_075051.4 | |
CAS1 domain-containing protein 1 | |
Research Use Only | |
CASD1 | |
Recombinant Protein | |
E. coli | |
ASRRYRGNDS CEYLLSSGRF LGEKVWQPHS CMMHKYKISE AKNCLVDKHI AFIGDSRIRQ LFYSFVKIIN PQFKEEGNKH ENIPFEDKTA SVKVDFLWHP EVNGSMKQCI KVWTEDSIAK PHVIVAGAAT WSIKIHNGSS EALSQYKMNI TSIAPLLEKL AKTSDVYWVL QDPVYEDLLS ENRKMITNEK IDAYNEAAVS ILNSSTRNSK SNVKMFSVSK LIAQETIMES LDGLHLPESS RETTAMILMN VYCNKILKPV DGSCCQPRPP VTLIQ | |
Greater than 90% as determined by SDS-PAGE. |
64921 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |