missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Caspase-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Caspase-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5762-ec-10ug
Detalles adicionales : Weight : 0.01000kg
Especificaciones
NP_001215.1 | |
Caspase-2 Protein | |
Research Use Only | |
CASP2 | |
Recombinant Protein | |
E. coli | |
AAPSAGSWST FQHKELMAAD RGRRILGVCG MHPHHQETLK KNRVVLAKQL LLSELLEHLL EKDIITLEMR ELIQAKVGSF SQNVELLNLL PKRGPQAFDA FCEALRETKQ GHLEDMLLTT LSGLQHVLPP LSCDYDLSLP FPVCESCPLY KKLRLSTDTV EHSLDNKDGP VCLQVKPCTP EFYQTHFQLA YRLQSRPRGL ALVLSNVHFT GEKELEFRSG GDVDHSTLVT LFKLLGYDVH VLCDQTAQEM QEKLQNFAQL PAHRVTDSCI VALLSHGVEG AIYGVDGKLL QLQEVFQLFD NANCPSLQNK PKMFFIQACR GDETDRGVDQ QDGKNHAGSP GCEESDAGKE KLPKMRLPTR SDMICGYACL KGTAAMRNTK RGSWYIEALA QVFSERACDM HVADMLVKVN ALIKDREGYA PGTEFHRCKE MSEYCSTLCR HLYLFPGHPP T | |
Greater than 90% as determined by SDS-PAGE. |
835 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |