missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coactosin-like Protein
A DNA sequence encoding the Homo sapiens (Human) Coactosin-like protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7594-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_066972.1 | |
Coactosin-like Protein | |
Research Use Only | |
COTL1 | |
Recombinant Protein | |
E. coli | |
ATKIDKEACR AAYNLVRDDG SAVIWVTFKY DGSTIVPGEQ GAEYQHFIQQ CTDDVRLFAF VRFTTGDAMS KRSKFALITW IGENVSGLQR AKTGTDKTLV KEVVQNFAKE FVISDRKELE EDFIKSELKK AGGANYDAQT E | |
Greater than 90% as determined by SDS-PAGE. |
23406 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |