missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCXR / HCR2 Protein
A DNA sequence encoding the Homo sapiens (Human) DCXR / HCR2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7792-ec-50ug
Weitere Details : Weight : 0.01000kg
Spezifikation
NP_001182147.1 | |
DCXR / HCR2 Protein | |
Research Use Only | |
DCXR | |
Recombinant Protein | |
E. coli | |
MELFLAGRRV LVTGAGKGIG RGTVQALHAT GARVVAVSRT QADLDSLVRE CPGIEPVCVD LGDWEATERA LGSVGPVDLL VNNAAVALLQ PFLEVTKEAF DRSFEVNLRA VIQVSQIVAR GLIARGVPGA IVNVSSQCSQ RAVTNHSVYC STKGALDMLT KVMALELGPH KIRVNAVNPT VVMTSMGQAT WSDPHKAKTM LNRIPLGKFA EVEHVVNAIL FLLSDRSGMT TGSTLPVEGG FWAC | |
Greater than 80% as determined by SDS-PAGE. |
51181 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |