missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Death effector domain-containing Protein
A DNA sequence encoding the Homo sapiens (Human) Death effector domain-containing protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5923-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001034800.1 | |
Death effector domain-containing Protein | |
Research Use Only | |
DEDD | |
Recombinant Protein | |
E. coli | |
MAGLKRRASQ VWPEEHGEQE HGLYSLHRMF DIVGTHLTHR DVRVLSFLFV DVIDDHERGL IRNGRDFLLA LERQGRCDES NFRQVLQLLR IITRHDLLPY VTLKRRRAVC PDLVDKYLEE TSIRYVTPRA LSDPEPRPPQ PSKTVPPHYP VVCCPTSGPQ MCSKRPARGR ATLGSQRKRR KSVTPDPKEK QTCDIRLRVR AEYCQHETAL QGNVFSNKQD PLERQFERFN QANTILKSRD LGSIICDIKF SELTYLDAFW RDYINGSLLE ALKGVFITDS LKQAVGHEAI KLLVNVDEED YELGRQKLLR NLMLQALP | |
Greater than 80% as determined by SDS-PAGE. |
9191 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |