missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA repair protein XRCC4 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA repair protein XRCC4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7540-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001304941.1 | |
DNA repair protein XRCC4 Protein | |
Research Use Only | |
XRCC4 | |
Recombinant Protein | |
E. coli | |
MERKISRIHL VSEPSITHFL QVSWEKTLES GFVITLTDGH SAWTGTVSES EISQEADDMA MEKGKYVGEL RKALLSGAGP ADVYTFNFSK ESCYFFFEKN LKDVSFRLGS FNLEKVENPA EVIRELICYC LDTIAENQAK NEHLQKENER LLRDWNDVQG RFEKCVSAKE ALETDLYKRF ILVLNEKKTK IRSLHNKLLN AAQEREKDIK QEGETAICSE MTADRDPVYD ESTDEESENQ TDLSGLASAA VSKDDSIISS LDVTDIAPSR KRRQRMQRNL GTEPKMAPQE NQLQEKENSR PDSSLPETSK KEHISAENMS LETLRNSSPE DLFDEI | |
Greater than 80% as determined by SDS-PAGE. |
7518 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |