missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DRAXIN Protein
A DNA sequence encoding the Homo sapiens (Human) DRAXIN, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7855-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_940947.3 | |
DRAXIN Protein | |
Research Use Only | |
DRAXIN | |
Recombinant Protein | |
E. coli | |
GALAPGTPAR NLPENHIDLP GPALWTPQAS HHRRRGPGKK EWGPGLPSQA QDGAVVTATR QASRLPEAEG LLPEQSPAGL LQDKDLLLGL ALPYPEKENR PPGWERTRKR SREHKRRRDR LRLHQGRALV RGPSSLMKKA ELSEAQVLDA AMEESSTSLA PTMFFLTTFE AAPATEESLI LPVTSLRPQQ AQPRSDGEVM PTLDMALFDW TDYEDLKPDG WPSAKKKEKH RGKLSSDGNE TSPAEGEPCD HHQDCLPGTC CDLREHLCTP HNRGLNNKCF DDCMCVEGLR CYAKFHRNRR VTRRKGRCVE PETANGDQGS FINV | |
Greater than 90% as determined by SDS-PAGE. |
374946 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |