missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DRAXIN Protein

A DNA sequence encoding the Homo sapiens (Human) DRAXIN, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7855-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16071201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_940947.3
DRAXIN Protein
Research Use Only
DRAXIN
Recombinant Protein
E. coli
GALAPGTPAR NLPENHIDLP GPALWTPQAS HHRRRGPGKK EWGPGLPSQA QDGAVVTATR QASRLPEAEG LLPEQSPAGL LQDKDLLLGL ALPYPEKENR PPGWERTRKR SREHKRRRDR LRLHQGRALV RGPSSLMKKA ELSEAQVLDA AMEESSTSLA PTMFFLTTFE AAPATEESLI LPVTSLRPQQ AQPRSDGEVM PTLDMALFDW TDYEDLKPDG WPSAKKKEKH RGKLSSDGNE TSPAEGEPCD HHQDCLPGTC CDLREHLCTP HNRGLNNKCF DDCMCVEGLR CYAKFHRNRR VTRRKGRCVE PETANGDQGS FINV
Greater than 90% as determined by SDS-PAGE.
374946
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers