missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Endothelin-1 receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Endothelin-1 receptor, was expressed in the hosts and tags indicated.
€ 590.00 - € 1910.00
Specifications
Accession Number | NP_001159527.1 |
---|---|
Gene ID (Entrez) | 1909 |
Name | Endothelin-1 receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019081
|
enQuireBio™
QP9952-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029081
|
enQuireBio™
QP9952-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009081
|
enQuireBio™
QP9952-IV-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001159527.1 | |
Endothelin-1 receptor Protein | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1909 | |
Research Use Only | |
EDNRA | |
Recombinant Protein | |
E. coli | |
MASMTGGQQM GRGSDNPERY STNLSNHVDD FTTFRGTELS FLVTTHQPTN LVLPSNGSMH NYCPQQTKIT SAFKYINTVI SCTIFIVGMV GNATLLRIIY QNKCMRNGHN ALIASLALGD LIYVVIDLPI NVFKLLAGRW PFDHNDFGVF LCKLFPFLQK SSVGITVLNL CALSVDRYRA VASWSRVQGI GIPLVTAIEI VSIWILSFIL AIPEAIGFVM VPFEYRGEQH KTCMLNATSK FMEFYQDVKD WWLFGFYFCM PLVCTAIFYT LMTCEMLNRR NGSLRIALSE HLKQRREVAK TVFCLVVIFA LCWFPVHLSR ILKKTVYNEM DKNRCELLSF LLLMDYIGIN LATMNSCINP IALYFVSKKF KNCFQSCLCC CCYQSKSLMT SVPMNGTSIQ WKDHDQNNHN TDRSSHKDSM N | |
Greater than 85% as determined by SDS-PAGE. |