missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FIBP Protein
A DNA sequence encoding the Homo sapiens (Human) FIBP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6048-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004205.2 | |
FIBP Protein | |
Research Use Only | |
FIBP | |
Recombinant Protein | |
E. coli | |
TSELDIFVGN TTLIDEDVYR LWLDGYSVTD AVALRVRSGI LEQTGATAAV LQSDTMDHY RTFHMLERLL HAPPKLLHQL IFQIPPSRQA LLIERYYAFD EAFVREVLGK KLSKGTKKDL DDISTKTGI TLKSCRRQFD NFKRVFKVVE EMRGSLVDNI QQHFLLSDRL ARDYAAIVFF ANNRFETGK KKLQYLSFGD FAFCAELMIQ NWTLGAVDSQ MDDMDMDLDK EFLQDLKELK VLVADKDLL DLHKSLVCTA LRGKLGVFSE MEANFKNLSR GLVNVAAKLT HNKDVRDLFV DLVEKFVEP CRSDHWPLSD VRFFLNQYSA SVHSLDGFRH QALWDRYMGT LRGCLLRLYH D | |
Greater than 90% as determined by SDS-PAGE. |
9158 | |
500 μg | |
Not determined. | |
Full Length of Isoform Short | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |