missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDNF family receptor alpha-like Protein
A DNA sequence encoding the Homo sapiens (Human) GDNF family receptor alpha-like, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7754-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_997293.2 | |
GDNF family receptor alpha-like Protein | |
Research Use Only | |
C6orf144 | |
Recombinant Protein | |
E. coli | |
SQTNNCTYLR EQCLRDANGC KHAWRVMEDA CNDSDPGDPC KMRNSSYCNL SIQYLVESNF QFKECLCTDD FYCTVNKLLG KKCINKSDNV KEDKFKWNLT TRSHHGFKGM WSCLEVAEAC VGDVVCNAQL ASYLKACSAN GNPCDLKQCQ AAIRFFYQNI PFNIAQMLAF CDCAQSDIPC QQSKEALHSK TCAVNMVPPP TCLSVIRSCQ NDELCRRHYR TFQSKCWQRV TRKCHEDENC ISTLSKQDLT CSGSDDCKAA YIDILGTVLQ VQCTCRTITQ SEESLCKIFQ HMLHRKSCFN YPTLSNVKGM ALYTRKHANK ITLTGFHSPF NGE | |
Greater than 90% as determined by SDS-PAGE. |
389400 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |