missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GIGYF2 Protein

A DNA sequence encoding the Homo sapiens (Human) GIGYF2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9814-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16098761

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001096616.1
GIGYF2 Protein
Research Use Only
GIGYF2
Recombinant Protein
Yeast
ERLASKLQEH RAKGVSIPLM HEAMQKWYYK DPQGEIQGPF NNQEMAEWFQ AGYFTMSLLV KRACDESFQP LGDIMKMWGR VPFSPGPAPP PHMGELDQER LTRQQELTAL YQMQHLQYQQ FLIQQQYAQV LAQQQKAALS SQQQQQLALL LQQFQTLKMR ISDQNIIPSV TRSVSVPDTG SIWELQPTAS QPTVWEGGSV WDLPLDTTTP GPALEQLQQL EKAKAAKLEQ ERREAEMRAK REEEERKRQE ELRRQQEEIL RRQQEEERKR REEEELARRK QEEALRRQRE QEIALRRQRE EEERQQQEEA LRRLEERRRE EEERRKQEEL LRKQEEEAAK WAREEEEAQR RLEENRLRME EEAARLRHEE EERKRKELEV QRQKELMRQR QQQQEALRRL QQQQQQQQLA QMKLPSSSTW GQQSNTTACQ SQATLSLAEI QKLEEERERQ LREEQRRQQR ELMKALQQQQ QQQQQKLSGW GNVSKPSGTT KSLLEIQQEE ARQMQKQQQQ QQQHQQPNRA R
Greater than 90% as determined by SDS-PAGE.
26058
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers