missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HAND2 Protein
A DNA sequence encoding the Homo sapiens (Human) HAND2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6142-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_068808.1 | |
HAND2 Protein | |
Research Use Only | |
HAND2 | |
Recombinant Protein | |
E. coli | |
MSLVGGFPHH PVVHHEGYPF AAAAAASRCS HEENPYFHGW LIGHPEMSPP DYSMALSYSP EYASGTANRK ERRRTQSINS AFAELRECIP NVPADTKLSK IKTLRLATSY IAYLMDLLAK DDQNGEAEAF KAEIKKTDVK EEKRKKELNE ILKSTVSSND KKTKGRTGWP QHVWALELKQ | |
Greater than 80% as determined by SDS-PAGE. |
9464 | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |