missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HAND2 Protein
A DNA sequence encoding the Homo sapiens (Human) HAND2, was expressed in the hosts and tags indicated.
€ 245.00 - € 3405.00
Specifications
Accession Number | NP_068808.1 |
---|---|
Gene ID (Entrez) | 9464 |
Name | HAND2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091110
|
enQuireBio™
QP6142-EC-100UG |
100 μg |
€ 770.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011120
|
enQuireBio™
QP6142-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001120
|
enQuireBio™
QP6142-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021120
|
enQuireBio™
QP6142-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031120
|
enQuireBio™
QP6142-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041120
|
enQuireBio™
QP6142-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_068808.1 | |
HAND2 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9464 | |
Research Use Only | |
HAND2 | |
Recombinant Protein | |
E. coli | |
MSLVGGFPHH PVVHHEGYPF AAAAAASRCS HEENPYFHGW LIGHPEMSPP DYSMALSYSP EYASGTANRK ERRRTQSINS AFAELRECIP NVPADTKLSK IKTLRLATSY IAYLMDLLAK DDQNGEAEAF KAEIKKTDVK EEKRKKELNE ILKSTVSSND KKTKGRTGWP QHVWALELKQ | |
Greater than 80% as determined by SDS-PAGE. |