missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein A1 Protein
A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6173-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002127.1 | |
Heterogeneous nuclear ribonucleoprotein A1 Protein | |
Research Use Only | |
HNRNPA1 | |
Recombinant Protein | |
E. coli | |
SKSESPKEPE QLRKLFIGGL SFETTDESLR SHFEQWGTLT DCVVMRDPNT KRSRGFGFVT YATVEEVDAA MNARPHKVDG RVVEPKRAVS REDSQRPGAH LTVKKIFVGG IKEDTEEHHL RDYFEQYGKI EVIEIMTDRG SGKKRGFAFV TFDDHDSVDK IVIQKYHTVN GHNCEVRKAL SKQEMASASS SQRGRSGSGN FGGGRGGGFG GNDNFGRGGN FSGRGGFGGS RGGGGYGGSG DGYNGFGNDG GYGGGGPGYS GGSRGYGSGG QGYGNQGSGY GGSGSYDSYN NGGGGGFGGG SGSNFGGGGS YNDFGNYNNQ SSNFGPMKGG NFGGRSSGPY GGGGQYFAKP RNQ | |
Greater than 90% as determined by SDS-PAGE. |
3178 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |