missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSP20 Protein
A cDNA sequence encoding the HSP20 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12326-10ug
Additional Details : Weight : 0.01000kg
Specifications
HSP20 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HSP20 lyophilized from 20mM Tris-acetate, pH 7.6, 10mM NaCl, 0.1mM EDTA, 0.1mM PMSF and 15mM b-mercaptoethanol. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |