missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hydroxymethylglutaryl-CoA lyase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Hydroxymethylglutaryl-CoA lyase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6168-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000182.2 | |
Hydroxymethylglutaryl-CoA lyase, mitochondrial Protein | |
Research Use Only | |
HMGCL | |
Recombinant Protein | |
E. coli | |
TLPKRVKIVE VGPRDGLQNE KNIVSTPVKI KLIDMLSEAG LSVIETTSFV SPKWVPQMGD HTEVLKGIQK FPGINYPVLT PNLKGFEAAV AAGAKEVVIF GAASELFTKK NINCSIEESF QRFDAILKAA QSANISVRGY VSCALGCPYE GKISPAKVAE VTKKFYSMGC YEISLGDTIG VGTPGIMKDM LSAVMQEVPL AALAVHCHDT YGQALANTLM ALQMGVSVVD SSVAGLGGCP YAQGASGNLA TEDLVYMLEG LGIHTGVNLQ KLLEAGNFIC QALNRKTSSK VAQATCKL | |
Greater than 80% as determined by SDS-PAGE. |
3155 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |