missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL2 / Interleukin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.
€ 202.00 - € 2155.00
Specifications
Accession Number | NP_000577.2 |
---|---|
Gene ID (Entrez) | 3558 |
Name | IL2 / Interleukin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091650
|
enQuireBio™
QP6216-YE-100UG |
100 μg |
€ 645.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041660
|
enQuireBio™
QP6216-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001660
|
enQuireBio™
QP6216-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021660
|
enQuireBio™
QP6216-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031660
|
enQuireBio™
QP6216-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011660
|
enQuireBio™
QP6216-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000577.2 | |
IL2 / Interleukin-2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3558 | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
Yeast | |
APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELKHLQCLE EELKPLEEVL NLAQSKNFHL RPRDLISNIN VIVLELKGSE TTFMCEYADE TATIVEFLNR WITFCQSIIS TLT | |
Greater than 90% as determined by SDS-PAGE. |