missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ILKAP Protein
A DNA sequence encoding the Homo sapiens (Human) ILKAP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6230-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_110395.1 | |
ILKAP Protein | |
Research Use Only | |
ILKAP | |
Recombinant Protein | |
E. coli | |
MDLFGDLPEP ERSPRPAAGK EAQKGPLLFD DLPPASSTDS GSGGPLLFDD LPPASSGDSG SLATSISQMV KTEGKGAKRK TSEEEKNGSE ELVEKKVCKA SSVIFGLKGY VAERKGEREE MQDAHVILND ITEECRPPSS LITRVSYFAV FDGHGGIRAS KFAAQNLHQN LIRKFPKGDV ISVEKTVKRC LLDTFKHTDE EFLKQASSQK PAWKDGSTAT CVLAVDNILY IANLGDSRAI LCRYNEESQK HAALSLSKEH NPTQYEERMR IQKAGGNVRD GRVLGVLEVS RSIGDGQYKR CGVTSVPDIR RCQLTPNDRF ILLACDGLFK VFTPEEAVNF ILSCLEDEKI QTREGKSAAD ARYEAACNRL ANKAVQRGSA DNVTVMVVRI GH | |
Greater than 90% as determined by SDS-PAGE. |
80895 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |