missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ILKAP Protein
A DNA sequence encoding the Homo sapiens (Human) ILKAP, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_110395.1 |
---|---|
Gene ID (Entrez) | 80895 |
Name | ILKAP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091770
|
enQuireBio™
QP6230-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011780
|
enQuireBio™
QP6230-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001780
|
enQuireBio™
QP6230-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021780
|
enQuireBio™
QP6230-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031780
|
enQuireBio™
QP6230-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041780
|
enQuireBio™
QP6230-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_110395.1 | |
ILKAP Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
80895 | |
Research Use Only | |
ILKAP | |
Recombinant Protein | |
E. coli | |
MDLFGDLPEP ERSPRPAAGK EAQKGPLLFD DLPPASSTDS GSGGPLLFDD LPPASSGDSG SLATSISQMV KTEGKGAKRK TSEEEKNGSE ELVEKKVCKA SSVIFGLKGY VAERKGEREE MQDAHVILND ITEECRPPSS LITRVSYFAV FDGHGGIRAS KFAAQNLHQN LIRKFPKGDV ISVEKTVKRC LLDTFKHTDE EFLKQASSQK PAWKDGSTAT CVLAVDNILY IANLGDSRAI LCRYNEESQK HAALSLSKEH NPTQYEERMR IQKAGGNVRD GRVLGVLEVS RSIGDGQYKR CGVTSVPDIR RCQLTPNDRF ILLACDGLFK VFTPEEAVNF ILSCLEDEKI QTREGKSAAD ARYEAACNRL ANKAVQRGSA DNVTVMVVRI GH | |
Greater than 90% as determined by SDS-PAGE. |