missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human INPP5A Protein

A DNA sequence encoding the Homo sapiens (Human) INPP5A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7598-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16019660

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001307971.1
INPP5A Protein
Research Use Only
INPP5A
Recombinant Protein
E. coli
MAGKAAAPGT AVLLVTANVG SLFDDPENLQ KNWLREFYQV VHTHKPHFMA LHCQEFGGKN YEASMSHVDK FVKELLSSDA MKEYNRARVY LDENYKSQEH FTALGSFYFL HESLKNIYQF DFKAKKYRKV AGKEIYSDTL ESTPMLEKEK FPQDYFPECK WSRKGFIRTR WCIADCAFDL VNIHLFHDAS NLVAWETSPS VYSGIRHKAL GYVLDRIIDQ RFEKVSYFVF GDFNFRLDSK SVVETLCTKA TMQTVRAADT NEVVKLIFRE SDNDRKVMLQ LEKKLFDYFN QEVFRDNNGT ALLEFDKELS VFKDRLYELD ISFPPSYPYS EDARQGEQYM NTRCPAWCDR ILMSPSAKEL VLRSESEEKV VTYDHIGPNV CMGDHKPVFL AFRIMPGAGK PHAHVHKCC
Greater than 90% as determined by SDS-PAGE.
3632
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers