missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KCNJ10 Protein

A DNA sequence encoding the Homo sapiens (Human) KCNJ10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9956-iv-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029091

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002232.2
KCNJ10 Protein
Research Use Only
KCNJ10
Recombinant Protein
E. coli
MTSVAKVYYS QTTQTESRPL MGPGIRRRRV LTKDGRSNVR MEHIADKRFL YLKDLWTTFI DMQWRYKLLL FSATFAGTWF LFGVVWYLVA VAHGDLLELD PPANHTPCVV QVHTLTGAFL FSLESQTTIG YGFRYISEEC PLAIVLLIAQ LVLTTILEIF ITGTFLAKIA RPKKRAETIR FSQHAVVASH NGKPCLMIRV ANMRKSLLIG CQVTGKLLQT HQTKEGENIR LNQVNVTFQV DTASDSPFLI LPLTFYHVVD ETSPLKDLPL RSGEGDFELV LILSGTVEST SATCQVRTSY LPEEILWGYE FTPAISLSAS GKYIADFSLF DQVVKVASPS GLRDSTVRYG DPEKLKLEES LREQAEKEGS ALSVRISNV
Greater than 90% as determined by SDS-PAGE.
3766
100 μg
Not determined.
Full Length Transmembrane Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers