missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCNJ10 Protein
A DNA sequence encoding the Homo sapiens (Human) KCNJ10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9956-iv-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002232.2 | |
KCNJ10 Protein | |
Research Use Only | |
KCNJ10 | |
Recombinant Protein | |
E. coli | |
MTSVAKVYYS QTTQTESRPL MGPGIRRRRV LTKDGRSNVR MEHIADKRFL YLKDLWTTFI DMQWRYKLLL FSATFAGTWF LFGVVWYLVA VAHGDLLELD PPANHTPCVV QVHTLTGAFL FSLESQTTIG YGFRYISEEC PLAIVLLIAQ LVLTTILEIF ITGTFLAKIA RPKKRAETIR FSQHAVVASH NGKPCLMIRV ANMRKSLLIG CQVTGKLLQT HQTKEGENIR LNQVNVTFQV DTASDSPFLI LPLTFYHVVD ETSPLKDLPL RSGEGDFELV LILSGTVEST SATCQVRTSY LPEEILWGYE FTPAISLSAS GKYIADFSLF DQVVKVASPS GLRDSTVRYG DPEKLKLEES LREQAEKEGS ALSVRISNV | |
Greater than 90% as determined by SDS-PAGE. |
3766 | |
10 μg | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |