missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lysyl oxidase homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Lysyl oxidase homolog 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6315-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005567.2 | |
Lysyl oxidase homolog 1 Protein | |
Research Use Only | |
Loxl1 | |
Recombinant Protein | |
E. coli | |
VGSDTVRGQA RHPFGFGQVP DNWREVAVGD STGMARARTS VSQQRHGGSA SSVSASAFAS TYRQQPSYPQ QFPYPQAPFV SQYENYDPAS RTYDQGFVYY RPAGGGVGAG AAAVASAGVI YPYQPRARYE EYGGGEELPE YPPQGFYPAP ERPYVPPPPP PPDGLDRRYS HSLYSEGTPG FEQAYPDPGP EAAQAHGGDP RLGWYPPYAN PPPEAYGPPR ALEPPYLPVR SSDTPPPGGE RNGAQQGRLS VGSVYRPNQN GRGLPDLVPD PNYVQASTYV QRAHLYSLRC AAEEKCLAST AYAPEATDYD VRVLLRFPQR VKNQGTADFL PNRPRHTWEW HSCHQHYHSM DEFSHYDLLD AATGKKVAEG HKASFCLEDS TCDFGNLKRY ACTSHTQGLS PGCYDTYNAD IDCQWIDITD VQPGNYILKV HVNPKYIVLE SDFTNNVVRC NIHYTGRYVS ATNCKIVQS | |
Greater than 90% as determined by SDS-PAGE. |
4016 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |