missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phospholipase D4 Protein
A DNA sequence encoding the Homo sapiens (Human) Phospholipase D4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7894-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_620145.2 | |
Phospholipase D4 Protein | |
Research Use Only | |
PLD4 | |
Recombinant Protein | |
E. coli | |
WQVPRPPTWG QVQPKDVPRS WEHGSSPAWE PLEAEARQQR DSCQLVLVES IPQDLPSAAG SPSAQPLGQA WLQLLDTAQE SVHVASYYWS LTGPDIGVND SSSQLGEALL QKLQQLLGRN ISLAVATSSP TLARTSTDLQ VLAARGAHVR QVPMGRLTRG VLHSKFWVVD GRHIYMGSAN MDWRSLTQVK ELGAVIYNCS HLAQDLEKTF QTYWVLGVPK AVLPKTWPQN FSSHFNRFQP FHGLFDGVPT TAYFSASPPA LCPQGRTRDL EALLAVMGSA QEFIYASVME YFPTTRFSHP PRYWPVLDNA LRAAAFGKGV RVRLLVGCGL NTDPTMFPYL RSLQALSNPA ANVSVDVKVF IVPVGNHSNI PFSRVNHSKF MVTEKAAYIG TSNWSEDYFS STAGVGLVVT QSPGAQPAGA TVQEQLRQLF ERDWSSRYAV GLDGQAPGQD CVWQG | |
Greater than 90% as determined by SDS-PAGE. |
122618 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |