missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PNRC2 Protein
A cDNA sequence encoding the PNRC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13087-5ug
Additional Details : Weight : 0.01000kg
Specifications
PNRC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMGGGERY NIPAPQSRNV SKNQQQLNRQ KTKEQNSQMK IVHKKKERGH GYNSSAAAWQ AMQNGGKNKN FPNNQSWNSS LSGPRLLFKS QANQNYAGAK FSEPPSPSVL PKPPSHWVPV SFNPSDKEIM TFQLKTLLKV QV | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PNRC2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |