missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein Hikeshi Protein

A DNA sequence encoding the Homo sapiens (Human) Protein Hikeshi, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7678-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020131

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_057485.2
Protein Hikeshi Protein
Research Use Only
C11orf73
Recombinant Protein
E. coli
MFGCLVAGRL VQTAAQQVAE DKFVFDLPDY ESINHVVVFM LGTIPFPEGM GGSVYFSYPD SNGMPVWQLL GFVTNGKPSA IFKISGLKSG EGSQHPFGAM NIVRTPSVAQ IGISVELLDS MAQQTPVGNA AVSSVDSFTQ FTQKMLDNFY NFASSFAVSQ AQMTPSPSEM FIPANVVLKW YENFQRRLAQ NPLFWKT
Greater than 90% as determined by SDS-PAGE.
51501
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers