missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTPN1 Protein

A cDNA sequence encoding the PTPN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10856-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935378

  • € 2395.00 / 1mg

Estimated Shipment: 03-02-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PTPN1 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MEMEKEFEQI DKSGSWAAIY QDIRHEASDF PCRVAKLPKN KNRNRYRDVS PFDHSRIKLHQEDNDYINAS LIKMEEAQRS YILTQGPLPN TCGHFWEMVW EQKSRGVVML NRVMEKGSLKCAQYWPQKEE KEMIFEDTNL KLTLISEDIK SYYTVRQLEL ENLTTQETRE ILHFHYTTWPDFGVPESPAS FLNFLFKVRE SGSLSPEHGP VVVHCSAGIG RSGTFCLADT CLLLMDKRKDPSSVDIKKVL LEMRKFRMGL IQTADQLRFS YLAVIEGAKF IMGDSSVQDQ WKELSHEDLE PPPEHIPPPPRPPKRILEPHN
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Specific activity is > 10,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C.
Human
Untagged
The protein (1 mg/ml) contains 25mM Tris-HCl, pH 7.5, 2mM beta-mercaptoethanol, 1mM EDTA, 1mM DTT and 20% Glycerol.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers