missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ptprn2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ptprn2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9873-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001295196.1 | |
Ptprn2 Protein | |
Research Use Only | |
Ptprn2 | |
Recombinant Protein | |
Yeast | |
AAPSSVPRGR QLPGRLGCLL EEGLCGASEA CVNDGVFGRC QKVPAMDFYR YEVSPVALQR LRVALQKLSG TGFTWQDDYT QYVMDQELAD LPKTYLRRPE ASSPARPSKH SVGSERRYSR EGGAALANAL RRHLPFLEAL SQAPASDVLA RTHTAQDRPP AEGDDRFSES ILTYVAHTSA LTYPPGSRTQ LREDLLPRTL GQLQPDELSP KVDSGVDRHH LMAALSAYAA QRPPAPPGEG SLEPQYLLRA PSRMPRPLLA PAAPQKWPSP LGDSEDPSST GDGARIHTLL KDLQRQPAEV RGLSGLELDG MAELMAGLMQ GVDHGVARGS PGRAALGESG EQADGPKATL RGDSFPDDGV QDDDDRLYQE VHRLSATLGG LLQDHGSRLL PGALPFARPL DMERKKSEHP ESSLSSEEET AGVENVKSQT YSKDLLGQQP HSEPGAAAFG ELQNQMPGPS KEEQSLPAGA QEALSDGLQL EVQPSEEEAR GYIVTDRDPL RPEEGRRLVE DVARLLQVPS SAFADVEVLG PAVTFKVSAN VQNVTTEDVE KATVDNKDKL EETSGLKILQ TGVGSKSKLK FLPPQAEQED STKF | |
Greater than 90% as determined by SDS-PAGE. |
5799 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |