missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RBBP9 Protein

A cDNA sequence encoding the RBBP9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13277-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953059

  • € 137.00 / 5µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RBBP9 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MASPSKAVIV PGNGGGDVTT HGWYGWVKKE LEKIPGFQCL AKNMPDPITA RESIWLPFME TELHCDEKTI IIGHSSGAIA AMRYAETHRV YAIVLVSAYT SDLGDENERA SGYFTRPWQW EKIKANCPYI VQFGSTDDPF LPWKEQQEVA DRLETKLHKF TDCGHFQNTE FHELITVVKS LLKVPA
Greater than 95% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RBBP9 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers