missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RLN3 Protein

A cDNA sequence encoding the RLN3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10859-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15915388

  • € 4090.00 / 1mg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RLN3 Protein
Research Use Only
The ED50, as measured by cAMP accumulation in human THP-1 cells, is less than 17.5ng/ml.
Human
Untagged
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
A chain: DVLAGLSSSC CKWGCSKSEI SSLC B chain: RAAPYGVRLCG REFIRAVIFT CGGSRW
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers