missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RLN3 Protein
A cDNA sequence encoding the RLN3 was constructed and used to recombinantly synthesize the protein.
€ 137.00 - € 4090.00
Specifications
Name | RLN3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50, as measured by cAMP accumulation in human THP-1 cells, is less than 17.5ng/ml. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935388
|
enQuireBio™
QP10859-5UG |
5 μg |
€ 137.00
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925388
|
enQuireBio™
QP10859-25UG |
25 μg |
€ 212.00
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915388
|
enQuireBio™
QP10859-1MG |
1 mg |
€ 4090.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RLN3 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
A chain: DVLAGLSSSC CKWGCSKSEI SSLC B chain: RAAPYGVRLCG REFIRAVIFT CGGSRW | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
The ED50, as measured by cAMP accumulation in human THP-1 cells, is less than 17.5ng/ml. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |