missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A15 / S100A7A Protein

A DNA sequence encoding the Homo sapiens (Human) S100A15 / S100A7A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7789-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16000811

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_789793.1
S100A15 / S100A7A Protein
Research Use Only
S100A7A
Recombinant Protein
E. coli
SNTQAERSII GMIDMFHKYT GRDGKIEKPS LLTMMKENFP NFLSACDKKG IHYLATVFEK KDKNEDKKID FSEFLSLLGD IAADYHKQSH GAAPCSGGSQ
Greater than 90% as determined by SDS-PAGE.
338324
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers