missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SERPINH1 Protein

A cDNA sequence encoding the SERPINH1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13469-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15953669

  • € 2725.00 / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SERPINH1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAAEVKKPAA AAAPGTAEKL SPKAATLAER SAGLAFSLYQ AMAKDQAVEN ILVSPVVVAS SLGLVSLGGK ATTASQAKAV LSAEQLRDEE VHAGLGELLR SLSNSTARNV TWKLGSRLYG PSSVSFADDF VRSSKQHYNC EHSKINFRDK RSALQSINEW AAQTTDGKLP EVTKDVERTD GALLVNAMFF KPHWDEKFHH KMVDNRGFMV TRSYTVGVMM MHRTGLYNYY DDEKEKLQIV EMPLAHKLSS LIILMPHHVE PLERLEKLLT KEQLKIWMGK MQKKAVAISL PKGVVEVTHD LQKHLAGLGL TEAIDKNKAD LSRMSGKKDL YLASVFHATA FELDTDGNPF DQDIYGREEL RSPKLFYADH PFIFLVRDTQ SGSLLFIGRL VRPKGDKMRD EL
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The SERPINH1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers