missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SUMO2 Protein

A cDNA sequence encoding the SUMO2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13649-GST-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944269

  • € 361.00 / 5µg

Estimated Shipment: 30-01-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

6613
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MADEKPKEGV KTENNDHINL KVAGQDGSVV QFKIKRHTPL SKLMKAYCER QGLSMRQIRF RFDGQPINET DTPAQLEMED EDTIDVFQQQ TGGVY
SUMO2 Protein
Research Use Only
SUMO2
Human
GST
SUMO2 at 100microg/ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers