missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transmembrane protein 65 Protein

A DNA sequence encoding the Homo sapiens(Human) Transmembrane protein 65, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9989-iv-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049151

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_919267.2
Transmembrane protein 65 Protein
Research Use Only
TMEM65
Recombinant Protein
E. coli
MEALNTAQGA RDFIYSLHST ERSCLLKELH RFESIAIAQE KLEAPPPTPG QLRYVFIHNA IPFIGFGFLD NAIMIVAGTH IEMSIGIILG ISTMAAAALG NLVSDLAGLG LAGYVEALAS RLGLSIPDLT PKQVDMWQTR LSTHLGKAVG VTIGCILGMF PLIFFGGGEE DEKLETKS
Greater than 85% as determined by SDS-PAGE.
157378
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers