missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza Matrix protein 1
A DNA sequence encoding the Influenza A virus (strain A/USA:Phila/1935 H1N1) Matrix protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9964-iv-100ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza Matrix protein 1 | |
Research Use Only | |
M2 | |
Recombinant Protein | |
E. coli | |
MSLLTEVETP IRNEWGCRCN GSSDPLVIAA SIIGILHLIL WILDRLLFKC IYRRFKYGLK RGPSTEGVPE SMREEYRKEQ QSAVDADDGH FVNIEPE | |
Greater than 80% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length Transmembrane Protein | |
Influenza | |
His | |
Tris-based buffer, 50% glycerol |