missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant M. tuberculosis Low molecular weight antigen MTB12 Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Low molecular weight antigen MTB12, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9684-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
WP_003917622.1 | |
100 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His | |
Tris-based buffer, 50% glycerol |
M. tuberculosis Low molecular weight antigen MTB12 Protein | |
Research Use Only | |
mtb12 | |
Recombinant Protein | |
Yeast | |
DPASAPDVPT AAQLTSLLNS LADPNVSFAN KGSLVEGGIG GTEARIADHK LKKAAEHGDL PLSFSVTNIQ PAAAGSATAD VSVSGPKLSS PVTQNVTFVN QGGWMLSRAS AMELLQAAGN | |
Greater than 90% as determined by SDS-PAGE. |