missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Macaque CCL20 / MIP-3 alpha Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5772-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001028026.1 | |
CCL20 / MIP-3 alpha Protein | |
Research Use Only | |
CCL20 | |
Recombinant Protein | |
E. coli | |
ASNFDCCLRY TDRILHPKFI VGFTQQLANE TCDINAVVFH TKKGLSVCAN PKQTWVKLIV RRLSKKINKM | |
Greater than 90% as determined by SDS-PAGE. |
574182 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Macaque | |
His-GST | |
Tris-based buffer, 50% glycerol |