missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Adipocyte enhancer-binding protein 1
A DNA sequence encoding the Mus musculus (Mouse) Adipocyte enhancer-binding protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7706-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001278786.1 | |
Adipocyte enhancer-binding protein 1 | |
Research Use Only | |
Aebp1 | |
Recombinant Protein | |
E. coli | |
QTVLTDDEIE EFLEGFLSEL ETQSPPREDD VEVQPLPEPT QRPRKSKAGG KQRADVEVPP EKNKDKEKKG KKDKGPKATK PLEGSTRPTK KPKEKPPKAT KKPKEKPPKA TKKPKEKPPK ATKKPKEKPP KATKRPSAGK KFSTVAPLET LDRLLPSPSN PSAQELPQKR DTPFPNAWQG QGEETQVEAK QPRPEPEEET EMPTLDYNDQ IEKEDYEDFE YIRRQKQPRP TPSRRRLWPE RPEEKTEEPE ERKEVEPPLK PLLPPDYGDS YVIPNYDDLD YYFPHPPPQK PDVGQEVDEE KEEMKKPKKE GSSPKEDTED KWTVEKNKDH KGPRKGEELE EEWAP | |
Greater than 80% as determined by SDS-PAGE. |
11568 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |