missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Adipocyte enhancer-binding protein 1
A DNA sequence encoding the Mus musculus (Mouse) Adipocyte enhancer-binding protein 1, was expressed in the hosts and tags indicated.
€ 309.00 - € 3405.00
Specifications
Accession Number | NP_001278786.1 |
---|---|
Gene ID (Entrez) | 11568 |
Name | Adipocyte enhancer-binding protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16080311
|
enQuireBio™
QP7706-EC-100UG |
100 μg |
€ 985.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000321
|
enQuireBio™
QP7706-EC-1MG |
1 mg |
€ 3405.00
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090311
|
enQuireBio™
QP7706-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010321
|
enQuireBio™
QP7706-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020321
|
enQuireBio™
QP7706-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030321
|
enQuireBio™
QP7706-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001278786.1 | |
Adipocyte enhancer-binding protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11568 | |
Research Use Only | |
Aebp1 | |
Recombinant Protein | |
E. coli | |
QTVLTDDEIE EFLEGFLSEL ETQSPPREDD VEVQPLPEPT QRPRKSKAGG KQRADVEVPP EKNKDKEKKG KKDKGPKATK PLEGSTRPTK KPKEKPPKAT KKPKEKPPKA TKKPKEKPPK ATKKPKEKPP KATKRPSAGK KFSTVAPLET LDRLLPSPSN PSAQELPQKR DTPFPNAWQG QGEETQVEAK QPRPEPEEET EMPTLDYNDQ IEKEDYEDFE YIRRQKQPRP TPSRRRLWPE RPEEKTEEPE ERKEVEPPLK PLLPPDYGDS YVIPNYDDLD YYFPHPPPQK PDVGQEVDEE KEEMKKPKKE GSSPKEDTED KWTVEKNKDH KGPRKGEELE EEWAP | |
Greater than 80% as determined by SDS-PAGE. |