missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Adipocyte enhancer-binding protein 1

A DNA sequence encoding the Mus musculus (Mouse) Adipocyte enhancer-binding protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7706-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030321

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001278786.1
Adipocyte enhancer-binding protein 1
Research Use Only
Aebp1
Recombinant Protein
E. coli
QTVLTDDEIE EFLEGFLSEL ETQSPPREDD VEVQPLPEPT QRPRKSKAGG KQRADVEVPP EKNKDKEKKG KKDKGPKATK PLEGSTRPTK KPKEKPPKAT KKPKEKPPKA TKKPKEKPPK ATKKPKEKPP KATKRPSAGK KFSTVAPLET LDRLLPSPSN PSAQELPQKR DTPFPNAWQG QGEETQVEAK QPRPEPEEET EMPTLDYNDQ IEKEDYEDFE YIRRQKQPRP TPSRRRLWPE RPEEKTEEPE ERKEVEPPLK PLLPPDYGDS YVIPNYDDLD YYFPHPPPQK PDVGQEVDEE KEEMKKPKKE GSSPKEDTED KWTVEKNKDH KGPRKGEELE EEWAP
Greater than 80% as determined by SDS-PAGE.
11568
50 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers