missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Alanine aminotransferase 1 Protein

A DNA sequence encoding the Mus musculus (Mouse) Alanine aminotransferase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9872-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068881

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_877957.1
Alanine aminotransferase 1 Protein
Research Use Only
GPT
Recombinant Protein
Yeast
ASQRNDRIQA SRNGLKGKVL TLDTMNPCVR RVEYAVRGPI VQRALELEQE LRQGVKKPFT EVIRANIGDA QAMGQRPITF FRQVLALCVY PNLLSSPDFP EDAKRRAERI LQACGGHSLG AYSISSGIQP IREDVAQYIE RRDGGIPADP NNIFLSTGAS DAIVTMLKLL VAGEGRARTG VLIPIPQYPL YSAALAELDA VQVDYYLDEE RAWALDIAEL RRALCQARDR CCPRVLCVIN PGNPTGQVQT RECIEAVIRF AFEEGLFLMA DEVYQDNVYA EGSQFHSFKK VLTEMGPPYA TQQELASFHS VSKGYMGECG FRGGYVEVVN MDAEVQKQMA KLMSVRLCPP VPGQALMGMV VSPPTPSEPS FKQFQAERQE VLAELAAKAK LTEQVFNEAP GIRCNPVQGA MYSFPQIQLP LKAVQRAQDL GLAPDMFFCL CLLEETGICV VPGSGFGQQE GTYHFRMTIL PPMEKLRVLL EKLRHFHAKF THEYS
Greater than 90% as determined by SDS-PAGE.
76282
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers