missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Bax Protein
A cDNA sequence encoding the Bax was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11131-2ug
Additional Details : Weight : 0.01000kg
Specifications
Bax Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDGSGEQLGS GGPTSSEQIM KTGAFLLQGF IQDRAGRMAG ETPELTLEQP PQDASTKKLS ECLRRIGDEL DSNMELQRMI ADVDTDSPRE VFFRVAADMF ADGNFNWGRV VALFYFASKL VLKALCTKVP ELIRTIMGWT LDFLRERLLV WIQDQGGWEG LLSYFGTPTW QTVTIFVAGV LTASLTIWKK MG | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
The protein contains 10mM Tris-HCL pH 8.0, 1mM EDTA and 250mM NaCl. |