missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Erythroferrone Protein
A DNA sequence encoding the Mus musculus (Mouse) Erythroferrone, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9819-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NC_000002.12 | |
Erythroferrone Protein | |
Research Use Only | |
ERFE | |
Recombinant Protein | |
Yeast | |
LGVPESAEPV GTHARPQPPG AELPAPPANS PPEPTIAHAH SVDPRDAWML FVKQSDKGIN SKRRSKARRL KLGLPGPPGP PGPQGPPGPF IPSEVLLKEF QLLLKGAVRQ RESHLEHCTR DLTTPASGSP SRVPAAQELD SQDPGALLAL LAATLAQGPR APRVEAAFHC RLRRDVQVDR RALHELGIYY LPEVEGAFHR GPGLNLTSGQ YTAPVAGFYA LAATLHVALT EQPRKGPTRP RDRLRLLICI QSLCQHNASL ETVMGLENSS ELFTISVNGV LYLQAGHYTS VFLDNASGSS LTVRSGSHFS AILLGL | |
Greater than 90% as determined by SDS-PAGE. |
151176 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |