missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Erythroferrone Protein
A DNA sequence encoding the Mus musculus (Mouse) Erythroferrone, was expressed in the hosts and tags indicated.
€ 245.00 - € 2645.00
Specifications
Accession Number | NC_000002.12 |
---|---|
Gene ID (Entrez) | 151176 |
Name | Erythroferrone Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008781
|
enQuireBio™
QP9819-YE-100UG |
100 μg |
€ 770.00
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048781
|
enQuireBio™
QP9819-YE-500UG |
500 μg |
€ 2195.00
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018781
|
enQuireBio™
QP9819-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038781
|
enQuireBio™
QP9819-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058781
|
enQuireBio™
QP9819-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028781
|
enQuireBio™
QP9819-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NC_000002.12 | |
Erythroferrone Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
151176 | |
Research Use Only | |
ERFE | |
Recombinant Protein | |
Yeast | |
LGVPESAEPV GTHARPQPPG AELPAPPANS PPEPTIAHAH SVDPRDAWML FVKQSDKGIN SKRRSKARRL KLGLPGPPGP PGPQGPPGPF IPSEVLLKEF QLLLKGAVRQ RESHLEHCTR DLTTPASGSP SRVPAAQELD SQDPGALLAL LAATLAQGPR APRVEAAFHC RLRRDVQVDR RALHELGIYY LPEVEGAFHR GPGLNLTSGQ YTAPVAGFYA LAATLHVALT EQPRKGPTRP RDRLRLLICI QSLCQHNASL ETVMGLENSS ELFTISVNGV LYLQAGHYTS VFLDNASGSS LTVRSGSHFS AILLGL | |
Greater than 90% as determined by SDS-PAGE. |