missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Galactocerebrosidase Protein
A DNA sequence encoding the Mus musculus (Mouse) Galactocerebrosidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9977-iv-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032105.2 | |
Galactocerebrosidase Protein | |
Research Use Only | |
GALC | |
Recombinant Protein | |
E. coli | |
YVLDDSDGLG REFDGIGAVS GGGATSRLLV NYPEPYRSEI LDYLFKPNFG ASLHILKVEI GGDGQTTDGT EPSHMHYELD ENYFRGYEWW LMKEAKKRNP DIILMGLPWS FPGWLGKGFS WPYVNLQLTA YYVVRWILGA KHYHDLDIDY IGIWNERPFD ANYIKELRKM LDYQGLQRVR IIASDNLWEP ISSSLLLDQE LWKVVDVIGA HYPGTYTVWN AKMSGKKLWS SEDFSTINSN VGAGCWSRIL NQNYINGNMT STIAWNLVAS YYEELPYGRS GLMTAQEPWS GHYVVASPIW VSAHTTQFTQ PGWYYLKTVG HLEKGGSYVA LTDGLGNLTI IIETMSHQHS MCIRPYLPYY NVSHQLATFT LKGSLREIQE LQVWYTKLGT PQQRLHFKQL DTLWLLDGSG SFTLELEEDE IFTLTTLTTG RKGSYPPPPS SKPFPTNYKD DFNVEYPLFS EAPNFADQTG VFEYYMNNED REHRFTLRQV LNQRPITWAA DASSTISVIG DHHWTNMTVQ CDVYIETPRS GGVFIAGRVN KGGILIRSAT GVFFWIFANG SYRVTADLGG WITYASGHAD VTAKRWYTLT LGIKGYFAFG MLNGTILWKN VRVKYPGHGW AAIGTHTFEF AQFDNFRVEA AR | |
Greater than 85% as determined by SDS-PAGE. |
14420 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
Untagged | |
Tris-based buffer, 50% glycerol |