missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Il27 alpha Protein
Recombinant Protein
Brand: enQuireBio™ QP5457-2ug
Additional Details : Weight : 0.01000kg
Specifications
NP_663611.1 | |
Il27 alpha Protein | |
Research Use Only | |
IL27Ra | |
Mouse | |
Untagged | |
10 mM sodium bicarbonate, pH 8.5 |
246779 | |
2 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MALVALSQPR VQCHASRYPV AVDCSWTPLQ APNSTRSTSF IATYRLGVAT QQQSQPCLQR SPQASRCTIP DVHLFSTVPY MLNVTAVHPG GASSSLLAFV AERIIKPDPP EGVRLRTAGQ RLQVLWHPPA SWPFPDIFSL KYRLRYRRRG ASHFRQVGPI EATTFTLRNS KPHAKYCIQV SAQDLTDYGK PSDWSLPGQV ESAPHKP | |
Greater than 95% as determined by SDS-PAGE. |