missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Thyroid hormone-inducible hepatic Protein
A DNA sequence encoding the Mus musculus (Mouse) Thyroid hormone-inducible hepatic protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7714-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_033407.1 | |
Thyroid hormone-inducible hepatic Protein | |
Research Use Only | |
Thrsp | |
Recombinant Protein | |
E. coli | |
MQVLTKRYPK NCLLTVMDRY SAVVRNMEQV VMIPSLLRDV QLSGPGGSVQ DGAPDLYTYF TMLKSICVEV DHGLLPREEW QAKVAGNETS EAENDAAETE EAEEDRISEE LDLEAQFHLH FCSLHHILTH LTRKAQEVTR KYQEMTGQVL | |
Greater than 90% as determined by SDS-PAGE. |
21835 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |